Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252197.1 | NP_001239126.1 | 2 | 572 | ||
NM_005853.5 | NP_005844.4 | 2 | 572 | |||
XM_011522809.1 | XP_011521111.1 | 2 | 1015 | |||
GenBank mRNA | AY335945.1 | - | 2 | 572 | ||
U90304.1 | - | 1 | 749 | |||
U90309.1 | - | 1 | 324 |