Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271629.1 | NP_001258558.1 | 3 | 414 | ||
NM_005920.3 | NP_005911.1 | 3 | 695 | |||
XM_005245169.4 | XP_005245226.1 | 3 | 677 | |||
XM_005245170.3 | XP_005245227.1 | 3 | 702 | |||
XM_006711330.3 | XP_006711393.1 | 3 | 406 | |||
XM_006711332.3 | XP_006711395.1 | 3 | 693 | |||
XM_006711333.2 | XP_006711396.1 | 3 | 690 | |||
XM_006711334.3 | XP_006711397.1 | 3 | 691 | |||
XM_011509569.2 | XP_011507871.1 | 3 | 727 | |||
XM_017001314.1 | XP_016856803.1 | 3 | 832 | |||
XM_017001315.1 | XP_016856804.1 | 3 | 427 | |||
GenBank mRNA | AB210005.1 | - | 3 | 609 | ||
AK308641.1 | - | 3 | 595 | |||
BC032479.1 | - | 3 | 606 | |||
BC040949.1 | - | 3 | 414 | 6044434 | ||
BC054520.1 | - | 3 | 590 | 6142751 | ||
L16794.1 | - | 3 | 395 |