Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077186.1 | NP_001070654.1 | 41 | 5752 | ||
NM_001145809.1 | NP_001139281.1 | 42 | 5851 | |||
NM_024729.3 | NP_079005.3 | 40 | 5728 | |||
XM_006723386.3 | XP_006723449.1 | 41 | 5813 | |||
XM_011527320.2 | XP_011525622.1 | 41 | 5830 | |||
XM_011527321.2 | XP_011525623.1 | 40 | 5806 | |||
XM_011527323.2 | XP_011525625.1 | 41 | 5820 | |||
GenBank mRNA | AB111886.1 | - | 31 | 4410 | ||
AB290169.1 | - | 42 | 5821 | |||
AK023943.1 | - | 10 | 1392 | |||
AK091982.1 | - | 1 | 701 | |||
AK122704.1 | - | 9 | 2979 | |||
AK303704.1 | - | 11 | 1678 | |||
AY165122.1 | - | 41 | 5728 | |||
AY203926.1 | - | 19 | 2793 | |||
BC000676.2 | - | 11 | 1146 | |||
BC007877.2 | - | 4 | 466 | |||
BC129803.1 | - | 29 | 3825 | |||
CR936653.1 | - | 11 | 1680 |