Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166237.1 | NP_001159709.1 | 6 | 944 | ||
NM_024736.6 | NP_079012.3 | 3 | 390 | |||
XM_011517301.1 | XP_011515603.1 | 3 | 359 | |||
GenBank mRNA | AF318341.1 | - | 4 | 428 | ||
AK022212.1 | - | 3 | 349 | |||
AK123985.1 | - | 2 | 573 | |||
AK127941.1 | - | 2 | 787 | |||
AK291817.1 | - | 6 | 944 | |||
AK303615.1 | - | 3 | 351 | |||
AY008304.1 | - | 3 | 379 | |||
BC008904.2 | - | 3 | 375 | 3634992 | ||
BC069000.1 | - | 3 | 330 | 6213121 | ||
DC398191.1 | - | 3 | 390 |