Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 920 | |||
NR_136193.1 | 4 | 1617 | ||||
NR_136194.1 | 4 | 1087 | ||||
NR_136197.1 | 4 | 991 | ||||
NR_136198.1 | 3 | 808 | ||||
NR_136199.1 | 5 | 1130 | ||||
NR_136200.1 | 4 | 1620 | ||||
NR_136201.1 | 4 | 1787 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1528 | ||
AK021601.1 | - | 1 | 1267 | |||
AK300846.1 | - | 4 | 998 | |||
BG519698.1 | - | 3 | 686 | |||
EU000853.1 | - | 4 | 833 | |||
EU000854.1 | - | 4 | 833 | |||
EU000855.1 | - | 3 | 1925 | |||
EU000856.1 | - | 3 | 1925 |