Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136474.2 | NP_001129946.1 | 7, 8 | 1100 | ||
NM_001136475.2 | NP_001129947.1 | 5, 6 | 816 | |||
NM_001301056.1 | NP_001287985.1 | 6, 7 | 1296 | |||
NM_024749.4 | NP_079025.2 | 4, 5 | 1164 | |||
XM_011509987.2 | XP_011508289.1 | 9, 10 | 1580 | |||
XM_011509988.2 | XP_011508290.1 | 10, 11 | 1654 | |||
XM_011509989.2 | XP_011508291.1 | 6, 7 | 1274 | |||
XM_011509991.2 | XP_011508293.2 | 5, 6 | 993 | |||
XM_011509995.1 | XP_011508297.1 | 4, 5 | 696 | |||
XM_017002351.1 | XP_016857840.1 | 7, 8 | 1414 | |||
XM_017002352.1 | XP_016857841.1 | 4, 5 | 847 | |||
GenBank mRNA | AK022567.1 | - | 7, 8 | 1026 | ||
AK096605.1 | - | 7, 8 | 964 | |||
AK290973.1 | - | 7, 8 | 1059 | |||
AK302675.1 | - | 5, 6 | 669 | |||
AY834202.1 | - | 5, 6 | 871 | |||
BC051856.1 | - | 4, 5 | 1049 | 6067012 |