Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031720.3 | NP_001026890.2 | 2 | 150 | ||
NM_024751.3 | NP_079027.2 | 2 | 332 | |||
XM_005263222.3 | XP_005263279.1 | 2 | 338 | |||
XM_011532248.2 | XP_011530550.1 | 2 | 294 | |||
XM_011532249.2 | XP_011530551.1 | 2 | 156 | |||
XM_011532252.2 | XP_011530554.1 | 2 | 528 | |||
GenBank mRNA | AK023335.1 | - | 2 | 329 | ||
AK225563.1 | - | 2 | 329 | |||
AK292355.1 | - | 2 | 153 | |||
BC032942.2 | - | 2 | 150 | 4838672 | ||
BX648355.1 | - | 2 | 333 |