Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024753.4 | NP_079029.3 | 25 | 3468 | ||
XM_011511871.2 | XP_011510173.1 | 20 | 6713 | |||
XM_011511872.2 | XP_011510174.1 | 25 | 3442 | |||
XM_017004967.1 | XP_016860456.1 | 25 | 3411 | |||
XM_017004968.1 | XP_016860457.1 | 20 | 4959 | |||
XM_017004969.1 | XP_016860458.1 | 23 | 3128 | |||
GenBank mRNA | AK096451.1 | - | 9 | 1095 | ||
AK292942.1 | - | 25 | 3411 | |||
BC035767.2 | - | 2 | 234 | |||
BC055424.1 | - | 25 | 3468 | 4837489 | ||
BC063579.1 | - | 7 | 812 | |||
BX647903.1 | - | 11 | 1219 |