Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024768.2 | NP_079044.2 | 6 | 1475 | ||
XM_005247781.3 | XP_005247838.1 | 6 | 1503 | |||
XM_011513161.2 | XP_011511463.1 | 6 | 935 | |||
XM_011513164.2 | XP_011511466.1 | 5 | 1548 | |||
GenBank mRNA | AK022115.1 | - | 1 | 998 | ||
AK022119.1 | - | 4 | 749 | |||
BC093800.1 | - | 2 | 186 | 7939645 | ||
BC112195.1 | - | 2 | 186 | 8327663 | ||
BX649124.1 | - | 1 | 4473 |