Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243797.1 | NP_001230726.1 | 3 | 187 | ||
NM_001243798.1 | NP_001230727.1 | 3 | 192 | |||
NM_006022.3 | NP_006013.1 | 3 | 529 | |||
NM_183422.3 | NP_904358.2 | 3 | 3511 | |||
GenBank mRNA | AB082525.1 | - | 3 | 3511 | ||
AJ222700.1 | - | 3 | 424 | |||
AK027071.1 | - | 3 | 2066 | |||
AK290751.1 | - | 3 | 464 | |||
AK312345.1 | - | 3 | 463 | |||
BC000456.2 | - | 3 | 445 | |||
BC016867.1 | - | 3 | 448 | |||
BC069207.1 | - | 3 | 437 | |||
BC105277.1 | - | 3 | 440 | |||
BC136475.1 | - | 3 | 3281 | 9020464 | ||
BC136482.1 | - | 3 | 3281 | 9020471 | ||
BC146664.1 | - | 3 | 444 | |||
BC146679.1 | - | 3 | 444 | |||
BF307401.1 | - | 3 | 189 | |||
BG703493.1 | - | 3 | 535 | |||
BI463370.1 | - | 3 | 195 | |||
CN344671.1 | - | 3 | 167 | |||
CR456711.1 | - | 3 | 233 | |||
CR627459.1 | - | 4 | 1704 | |||
D38585.1 | - | 3 | 233 | |||
D87061.1 | - | 3 | 233 | |||
U35048.1 | - | 3 | 415 |