Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039580.1 | NP_001034669.1 | 2, 3 | 347 | ||
XM_006714306.3 | XP_006714369.1 | 2, 3 | 495 | |||
XM_011532253.1 | XP_011530555.1 | 2, 3 | 550 | |||
XM_011532254.2 | XP_011530556.1 | 2, 3 | 459 | |||
XM_017008616.1 | XP_016864105.1 | 2, 3 | 550 | |||
XM_017008617.1 | XP_016864106.1 | 2, 3 | 367 | |||
XM_017008618.1 | XP_016864107.1 | 2, 3 | 325 | |||
GenBank mRNA | AK024812.1 | - | 2, 3 | 347 | ||
AK301075.1 | - | 2, 3 | 214 | |||
AY690636.1 | - | 2, 3 | 347 | |||
BC022852.1 | - | 2, 3 | 175 | |||
BC128540.1 | - | 2, 3 | 262 | |||
BC128541.1 | - | 2, 3 | 262 |