Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136041.2 | NP_001129513.1 | 7 | 561 | ||
NM_024827.3 | NP_079103.2 | 7 | 662 | |||
XM_005265491.1 | XP_005265548.1 | 7 | 627 | |||
XM_005265492.1 | XP_005265549.1 | 4 | 469 | |||
XM_011534131.1 | XP_011532433.1 | 7 | 750 | |||
XM_011534132.2 | XP_011532434.2 | 4 | 315 | |||
XM_011534135.1 | XP_011532437.1 | 6 | 469 | |||
GenBank mRNA | AK025426.1 | - | 7 | 553 | ||
AK025890.1 | - | 7 | 553 | |||
AK293223.1 | - | 7 | 494 | |||
AK299021.1 | - | 3 | 233 | |||
BC009676.1 | - | 6 | 538 | 3906049 | ||
CR457341.1 | - | 3 | 163 |