Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256378.1 | NP_001243307.1 | 2 | 418 | ||
NM_001256379.1 | NP_001243308.1 | 2 | 259 | |||
NM_024834.3 | NP_079110.1 | 2 | 418 | |||
XM_005270157.4 | XP_005270214.1 | 2 | 500 | |||
XM_011540170.1 | XP_011538472.1 | 2 | 259 | |||
XM_017016663.1 | XP_016872152.1 | 2 | 500 | |||
XM_017016664.1 | XP_016872153.1 | 2 | 259 | |||
GenBank mRNA | AK023143.1 | - | 2 | 289 | ||
AK094075.1 | - | 2 | 253 | |||
BC004183.2 | - | 2 | 384 | 2961492 | ||
BC007219.1 | - | 2 | 384 | 2961492 | ||
BQ929053.1 | - | 2 | 418 | |||
CR457299.1 | - | 2 | 119 | |||
DA381502.1 | - | 2 | 259 | |||
KU178788.1 | - | 2 | 119 | |||
KU178789.1 | - | 2 | 119 |