Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166237.1 | NP_001159709.1 | 14 | 2307 | ||
NM_024736.6 | NP_079012.3 | 11 | 1753 | |||
GenBank mRNA | AF318341.1 | - | 12 | 1792 | ||
BC008904.2 | - | 11 | 1738 | 3634992 | ||
BC069000.1 | - | 11 | 1693 | 6213121 |