Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHCBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324319.1 | NP_001311248.1 | 2 | 238 | ||
NM_024745.4 | NP_079021.3 | 2 | 235 | |||
NR_136738.1 | 2 | 238 | ||||
XM_011523335.2 | XP_011521637.1 | 2 | 3769 | |||
GenBank mRNA | AK055931.1 | - | 2 | 206 | ||
AK292164.1 | - | 2 | 202 | |||
AK310430.1 | - | 2 | 204 | |||
AK315706.1 | - | 2 | 204 | |||
BC030699.1 | - | 2 | 210 | 4829528 | ||
BP245197.1 | - | 2 | 235 |