Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024747.5 | NP_079023.2 | 1 | 2054 | ||
GenBank mRNA | AF536238.1 | - | 1 | 2019 | ||
AK026154.1 | - | 1 | 847 | |||
BC011594.1 | - | 1 | 2019 | 4578480 | ||
BC014993.1 | - | 1 | 2031 | 4869397 | ||
BC080564.1 | - | 2 | 604 |