Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031720.3 | NP_001026890.2 | 2 | 245 | ||
XM_005263222.3 | XP_005263279.1 | 2 | 433 | |||
XM_011532248.2 | XP_011530550.1 | 2 | 389 | |||
XM_011532249.2 | XP_011530551.1 | 2 | 251 | |||
XM_011532252.2 | XP_011530554.1 | 2 | 623 | |||
GenBank mRNA | AK023335.1 | - | 2 | 424 | ||
AK225563.1 | - | 2 | 424 | |||
AK292355.1 | - | 2 | 248 | |||
BC032942.2 | - | 2 | 245 | 4838672 |