Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024756.2 | NP_079032.2 | 5 | 657 | ||
XM_005270153.1 | XP_005270210.1 | 5 | 696 | |||
XM_006717970.3 | XP_006718033.1 | 5 | 2636 | |||
GenBank mRNA | AK023527.1 | - | 5 | 604 | ||
AK304727.1 | - | 5 | 514 | |||
AK308401.1 | - | 4 | 498 | |||
BC064415.1 | - | 5 | 612 | |||
BC072456.1 | - | 5 | 553 | |||
BC094744.1 | - | 5 | 604 | 30343159 |