Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000111.2 | NP_000102.1 | 3 | 366 | ||
XM_011515867.2 | XP_011514169.1 | 3 | 347 | |||
GenBank mRNA | BC025671.1 | - | 3 | 340 | 5202612 | |
BP262081.1 | - | 3 | 366 | |||
L02785.1 | - | 3 | 339 |