Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024372.1 | NP_001019543.1 | 2 | 503 | ||
NM_024812.2 | NP_079088.1 | 3 | 670 | |||
GenBank mRNA | AK022077.1 | - | 3 | 648 | ||
AK093819.1 | - | 2 | 356 | |||
AK310816.1 | - | 2 | 403 | |||
BC011517.1 | - | 3 | 612 | 4183341 | ||
BC035038.2 | - | 2 | 483 |