Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020946.1 | NP_065997.1 | 3, 4 | 358 | ||
NM_024820.2 | NP_079096.2 | 3, 4 | 358 | |||
XM_005252109.2 | XP_005252166.1 | 3, 4 | 207 | |||
XM_005252111.4 | XP_005252168.1 | 3, 4 | 1174 | |||
XM_005252113.4 | XP_005252170.1 | 3, 4 | 1163 | |||
XM_006717195.3 | XP_006717258.1 | 3, 4 | 1175 | |||
XM_011518882.2 | XP_011517184.1 | 3, 4 | 1177 | |||
XM_011518883.2 | XP_011517185.1 | 3, 4 | 1178 | |||
XM_011518885.2 | XP_011517187.1 | 2, 3 | 870 | |||
XM_011518886.2 | XP_011517188.1 | 3, 4 | 1175 | |||
XM_011518887.2 | XP_011517189.1 | 3, 4 | 1166 | |||
XM_017014948.1 | XP_016870437.1 | 3, 4 | 207 | |||
XM_017014949.1 | XP_016870438.1 | 3, 4 | 1178 | |||
XM_017014950.1 | XP_016870439.1 | 3, 4 | 1177 | |||
GenBank mRNA | AB046828.2 | - | 2, 3 | 292 | ||
AK024782.1 | - | 3, 4 | 346 | |||
AK225104.1 | - | 3, 4 | 346 | |||
BC039703.1 | - | 3, 4 | 211 | 4999484 |