Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167575.1 | NP_001161047.1 | 3, 4 | 863 | ||
NM_024828.3 | NP_079104.3 | 3, 4 | 709 | |||
GenBank mRNA | AK023719.1 | - | 3, 4 | 683 | ||
AK300739.1 | - | 3, 4 | 477 | |||
BC014658.1 | - | 3, 4 | 685 | 4648932 | ||
BC071953.1 | - | 3, 4 | 608 | 5431506 |