Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024831.7 | NP_079107.6 | 2 | 528 | ||
XM_006716485.2 | XP_006716548.1 | 2 | 314 | |||
XM_017014004.1 | XP_016869493.1 | 2 | 314 | |||
GenBank mRNA | AF286340.1 | - | 2 | 618 | ||
AF432215.1 | - | 2 | 276 | |||
AK314693.1 | - | 2 | 302 | |||
AY028423.1 | - | 2 | 312 | |||
BM853546.1 | - | 2 | 528 | |||
DB082992.1 | - | 2 | 316 |