Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319987.1 | NP_001306916.1 | 5 | 1770 | ||
NM_024832.4 | NP_079108.3 | 6 | 2023 | |||
XM_011537164.2 | XP_011535466.1 | 5 | 1758 | |||
XM_011537165.2 | XP_011535467.1 | 5 | 1958 | |||
XM_017021651.1 | XP_016877140.1 | 6 | 1905 | |||
XM_017021652.1 | XP_016877141.1 | 6 | 2031 | |||
XM_017021653.1 | XP_016877142.1 | 6 | 2031 | |||
XM_017021654.1 | XP_016877143.1 | 6 | 2031 | |||
GenBank mRNA | AB060338.1 | - | 1 | 569 | ||
AB081753.1 | - | 6 | 1871 | |||
AK026092.1 | - | 1 | 472 | |||
AK074176.1 | - | 1 | 420 | |||
AK090451.1 | - | 1 | 660 | |||
AK128338.1 | - | 5 | 1770 | |||
BC025248.1 | - | 1 | 892 | |||
BC070062.1 | - | 7 | 1783 | 30332851 | ||
BX248258.1 | - | 6 | 2009 | |||
CR936654.1 | - | 1 | 57 |