Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024835.4 | NP_079111.1 | 4 | 681 | ||
XM_005257689.2 | XP_005257746.1 | 4 | 657 | |||
XM_011525265.2 | XP_011523567.1 | 3 | 420 | |||
XM_011525266.1 | XP_011523568.1 | 4 | 657 | |||
XM_017025107.1 | XP_016880596.1 | 3 | 394 | |||
GenBank mRNA | AF126964.1 | - | 3 | 412 | ||
AF268387.1 | - | 4 | 601 | |||
AK024883.1 | - | 4 | 637 | |||
AK290687.1 | - | 3 | 437 | |||
AK292161.1 | - | 4 | 650 | |||
BC137491.1 | - | 4 | 547 | 9021491 | ||
BG393068.1 | - | 614 | ||||
DC424145.1 | - | 4 | 657 |