Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGMAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024758.4 | NP_079034.3 | 7 | 1486 | ||
GenBank mRNA | AK130034.1 | - | 7 | 1282 | ||
AY057097.1 | - | 7 | 1386 |