Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031855.2 | NP_001027026.1 | 4 | 1437 | ||
NM_001289109.1 | NP_001276038.1 | 5 | 1223 | |||
NM_024778.5 | NP_079054.3 | 3 | 1314 | |||
NR_110311.1 | 4 | 1437 | ||||
XM_005262476.2 | XP_005262533.1 | 5 | 1593 | |||
XM_011531398.2 | XP_011529700.1 | 5 | 1593 | |||
XM_017029845.1 | XP_016885334.1 | 3 | 1314 | |||
GenBank mRNA | AK026265.1 | - | 3 | 566 | ||
AK091777.1 | - | 4 | 1433 | |||
AK097641.1 | - | 5 | 1223 | |||
AK290198.1 | - | 3 | 1310 | |||
BC099847.1 | - | 4 | 1301 | 40007729 | ||
BC100671.1 | - | 4 | 1301 | 40007726 | ||
BC103491.1 | - | 3 | 1178 | 40007723 |