Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSPP1 SIRNA Assays ›
Gene Symbol:
CSPP1
Gene Name:
centrosome and spindle pole associated protein 1
Gene Aliases:
CSPP, JBTS21
Chromosome Location:
Chr.8: 67062417 - 67196614 on Build GRCh38
UniGene ID:
Hs.370147
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31762
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291339.1
NP_001278268.1
20
2864
NM_024790.6
NP_079066.5
23
2864
XM_005251305.3
XP_005251362.2
26
3348
XM_006716474.2
XP_006716537.2
25
3195
XM_006716477.2
XP_006716540.2
21
2817
XM_011517598.1
XP_011515900.1
27
3390
XM_011517599.1
XP_011515901.1
26
3366
XM_011517600.1
XP_011515902.1
25
3324
XM_011517601.1
XP_011515903.1
25
3285
XM_011517602.1
XP_011515904.1
24
3243
XM_011517603.1
XP_011515905.1
26
3073
XM_011517607.1
XP_011515909.1
25
2953
XM_011517608.1
XP_011515910.1
24
2950
XM_011517609.1
XP_011515911.1
23
3072
XM_011517611.2
XP_011515913.1
11
1449
XM_017013847.1
XP_016869336.1
25
3249
XM_017013848.1
XP_016869337.1
24
3225
XM_017013849.1
XP_016869338.1
24
3186
XM_017013850.1
XP_016869339.1
23
3144
XM_017013851.1
XP_016869340.1
23
2908
XM_017013852.1
XP_016869341.1
22
2991
XM_017013853.1
XP_016869342.1
22
2755
XM_017013854.1
XP_016869343.1
20
2793
XM_017013855.1
XP_016869344.1
18
2559
XM_017013856.1
XP_016869345.1
18
2377
XM_017013857.1
XP_016869346.1
19
2427
XM_017013858.1
XP_016869347.1
17
2175
XM_017013859.1
XP_016869348.1
13
1577
GenBank mRNA
AJ583433.1
-
20
2864
AK026143.1
-
311
AK308372.1
-
7
840
AM156947.1
-
23
2865
BC022867.1
-
5
363
BX649147.1
-
23
2866
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
positive regulation of cytokinesis
positive regulation of cell division
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline