Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320210.1 | NP_001307139.1 | 9 | 1066 | ||
NM_001321238.1 | NP_001308167.1 | 7 | 930 | |||
NM_001321239.1 | NP_001308168.1 | 7 | 1780 | |||
NM_024818.4 | NP_079094.1 | 9 | 1916 | |||
NM_198329.3 | NP_938143.1 | 9 | 939 | |||
GenBank mRNA | AB154406.1 | - | 9 | 875 | ||
AK026904.1 | - | 9 | 1109 | |||
AK027563.1 | - | 8 | 708 | |||
AK056238.1 | - | 9 | 1917 | |||
AK129616.1 | - | 10 | 975 | |||
AL136757.1 | - | 9 | 929 | |||
AL832669.1 | - | 5 | 2302 | |||
AY253672.1 | - | 9 | 1068 | |||
BC009737.1 | - | 9 | 1045 | 3879061 | ||
BC048015.1 | - | 9 | 1023 |