Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304797.1 | NP_001291726.1 | 4 | 741 | ||
NM_024821.3 | NP_079097.1 | 7 | 1080 | |||
XM_005261748.3 | XP_005261805.1 | 8 | 1170 | |||
GenBank mRNA | AK026002.1 | - | 7 | 1023 | ||
AK299311.1 | - | 4 | 684 | |||
BC017693.2 | - | 7 | 1005 | 4403280 |