Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024835.4 | NP_079111.1 | 6 | 896 | ||
XM_005257689.2 | XP_005257746.1 | 6 | 872 | |||
XM_011525265.2 | XP_011523567.1 | 5 | 635 | |||
XM_017025107.1 | XP_016880596.1 | 5 | 609 | |||
GenBank mRNA | AF126964.1 | - | 5 | 627 | ||
AF268387.1 | - | 6 | 816 | |||
AK024883.1 | - | 6 | 852 | |||
AK290687.1 | - | 5 | 652 | |||
AK292161.1 | - | 6 | 865 | |||
AK301614.1 | - | 3 | 184 | |||
BC137491.1 | - | 6 | 762 | 9021491 |