Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031721.3 | NP_001026891.2 | 5 | 659 | ||
NM_024840.3 | NP_079116.2 | 5 | 551 | |||
XM_005259269.3 | XP_005259326.1 | 4 | 765 | |||
XM_011527333.2 | XP_011525635.1 | 5 | 875 | |||
XM_017027315.1 | XP_016882804.1 | 4 | 723 | |||
GenBank mRNA | AK027565.1 | - | 5 | 551 | ||
BC057776.1 | - | 5 | 618 | 30338140 |