Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160167.1 | NP_001153639.1 | 5 | 710 | ||
NM_001160168.1 | NP_001153640.1 | 3 | 250 | |||
NM_001160169.1 | NP_001153641.1 | 4 | 446 | |||
NM_024841.4 | NP_079117.3 | 6 | 696 | |||
GenBank mRNA | AK024275.1 | - | 3 | 197 | ||
AK055042.1 | - | 5 | 597 | |||
BX640959.1 | - | 6 | 677 | |||
DA344861.1 | - | 4 | 548 | |||
DR001466.1 | - | 4 | 379 |