Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024898.3 | NP_079174.2 | 3, 4 | 247 | ||
XM_006722905.2 | XP_006722968.2 | 2, 3 | 117 | |||
XM_006722906.3 | XP_006722969.1 | 3, 4 | 176 | |||
XM_011528318.2 | XP_011526620.1 | 2, 3 | 167 | |||
GenBank mRNA | AK026410.1 | - | 3, 4 | 226 | ||
AK225623.1 | - | 3, 4 | 226 | |||
AK303509.1 | - | 2, 3 | 152 | |||
AL713770.1 | - | 2, 3 | 355 | |||
BC033437.1 | - | 3, 4 | 214 | 5187180 | ||
DC378383.1 | - | 3, 4 | 247 |