Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP8B4 SIRNA Assays ›
Gene Symbol:
ATP8B4
Gene Name:
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases:
ATPIM
Chromosome Location:
Chr.15: 49858238 - 50182096 on Build GRCh38
UniGene ID:
Hs.511311
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31899
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP8B4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024837.3
NP_079113.2
28
3465
NR_073596.1
28
3364
NR_073597.1
27
3317
NR_073598.1
29
3572
XM_011522046.2
XP_011520348.1
30
3854
XM_011522047.2
XP_011520349.1
29
4379
XM_011522048.1
XP_011520350.1
29
3652
XM_011522049.2
XP_011520351.1
29
3762
XM_011522051.2
XP_011520353.1
30
3689
XM_011522052.2
XP_011520354.1
29
3747
XM_011522053.1
XP_011520355.1
29
3528
XM_011522056.2
XP_011520358.2
29
3914
XM_011522058.2
XP_011520360.1
25
4123
XM_011522059.1
XP_011520361.1
24
3144
XM_011522060.1
XP_011520362.1
27
3379
XM_011522061.1
XP_011520363.1
26
3294
XM_011522062.1
XP_011520364.1
24
3133
XM_011522063.1
XP_011520365.1
24
3115
XM_011522064.1
XP_011520366.1
20
2816
XM_011522069.2
XP_011520371.1
19
2637
XM_011522070.1
XP_011520372.1
15
2057
XM_017022587.1
XP_016878076.1
28
3830
XM_017022588.1
XP_016878077.1
28
3755
XM_017022589.1
XP_016878078.1
28
3725
XM_017022590.1
XP_016878079.1
30
3666
XM_017022591.1
XP_016878080.1
29
3613
XM_017022592.1
XP_016878081.1
28
3676
XM_017022594.1
XP_016878083.1
25
3174
XM_017022595.1
XP_016878084.1
25
5179
XM_017022596.1
XP_016878085.1
20
2807
GenBank mRNA
AB075819.1
-
29
3572
AK025125.1
-
5
605
AK123747.1
-
9
1231
BC057236.1
-
4
557
BC144297.1
-
27
3313
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
phospholipid translocation
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline