Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292039.1 | NP_001278968.1 | 5 | 3295 | ||
NM_001292040.1 | NP_001278969.1 | 4 | 4335 | |||
NM_002747.3 | NP_002738.2 | 6 | 4711 | |||
XM_005258299.3 | XP_005258356.1 | 6 | 9073 | |||
XM_011526074.2 | XP_011524376.1 | 6 | 9255 | |||
XM_011526075.2 | XP_011524377.1 | 6 | 4713 | |||
XM_011526076.2 | XP_011524378.1 | 6 | 5076 | |||
XM_011526077.2 | XP_011524379.1 | 5 | 5722 | |||
XM_011526078.2 | XP_011524380.1 | 4 | 3827 | |||
XM_017025839.1 | XP_016881328.1 | 6 | 4644 | |||
GenBank mRNA | BC040493.1 | - | 4 | 4307 | ||
BC050299.1 | - | 6 | 4710 | 5303135 | ||
X59727.1 | - | 5 | 4199 |