Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024859.3 | NP_079135.3 | 4 | 412 | ||
XM_005278062.4 | XP_005278119.1 | 4 | 451 | |||
XM_005278063.4 | XP_005278120.1 | 4 | 450 | |||
XM_005278064.4 | XP_005278121.1 | 2 | 345 | |||
XM_005278065.3 | XP_005278122.1 | 2 | 433 | |||
XM_006724560.3 | XP_006724623.1 | 4 | 496 | |||
XM_011543985.2 | XP_011542287.1 | 3 | 566 | |||
XM_011543986.2 | XP_011542288.1 | 2 | 320 | |||
XM_011543987.2 | XP_011542289.1 | 2 | 341 | |||
GenBank mRNA | AK025340.1 | - | 2 | 267 | ||
AK094251.1 | - | 2 | 899 | |||
AK096058.1 | - | 2 | 1507 | |||
DA072017.1 | - | 3 | 194 | |||
DQ884401.1 | - | 1 | 188 |