Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026807.2 | 3 | 799 | |||
NR_121612.1 | 3 | 850 | ||||
NR_121613.1 | 2 | 784 | ||||
NR_121614.1 | 2 | 733 | ||||
GenBank mRNA | AI498652.1 | - | 2 | 255 | ||
AI808466.1 | - | 3 | 271 | |||
AI985403.1 | - | 3 | 372 | |||
AK023251.1 | - | 3 | 799 | |||
AW274058.1 | - | 3 | 305 | |||
BX118240.1 | - | 1 | 97 | |||
CD674415.1 | - | 2 | 300 |