Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNDIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323606.1 | NP_001310535.1 | 4 | 2040 | ||
NM_001323607.1 | NP_001310536.1 | 3 | 716 | |||
NM_024893.2 | NP_079169.1 | 3 | 1151 | |||
XM_011529347.2 | XP_011527649.1 | 3 | 969 | |||
XM_011529348.2 | XP_011527650.1 | 3 | 969 | |||
XM_011529349.2 | XP_011527651.1 | 3 | 969 | |||
XM_011529350.1 | XP_011527652.1 | 4 | 1605 | |||
XM_011529351.1 | XP_011527653.1 | 4 | 1512 | |||
XM_011529352.2 | XP_011527654.1 | 3 | 710 | |||
XM_011529353.2 | XP_011527655.1 | 3 | 727 | |||
XM_011529354.2 | XP_011527656.1 | 3 | 707 | |||
XM_011529356.2 | XP_011527658.1 | 3 | 969 | |||
XM_011529358.2 | XP_011527660.1 | 3 | 969 | |||
XM_017028064.1 | XP_016883553.1 | 3 | 3986 | |||
XM_017028065.1 | XP_016883554.1 | 4 | 1512 | |||
XM_017028066.1 | XP_016883555.1 | 3 | 869 | |||
XM_017028067.1 | XP_016883556.1 | 3 | 775 | |||
XM_017028068.1 | XP_016883557.1 | 3 | 1736 | |||
GenBank mRNA | AK024282.1 | - | 3 | 1151 | ||
BC030637.1 | - | 3 | 653 | 4821088 | ||
CR457325.1 | - | 2 | 518 |