Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAQR6 SIRNA Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
PRdelta
Chromosome Location:
Chr.1: 156243321 - 156248488 on Build GRCh38
UniGene ID:
Hs.235873
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31947
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAQR6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272104.1
NP_001259033.1
8
1735
NM_001272105.1
NP_001259034.1
9
1807
NM_001272106.1
NP_001259035.1
6
1701
NM_001272107.1
NP_001259036.1
7
1870
NM_001272108.1
NP_001259037.1
6
1360
NM_001272109.1
NP_001259038.1
5
1625
NM_001272110.1
NP_001259039.1
7
1608
NM_001272111.1
NP_001259040.1
7
1566
NM_001272112.1
NP_001259041.1
5
1393
NM_001272113.1
NP_001259042.1
5
1263
NM_024897.3
NP_079173.2
7
1941
NM_198406.2
NP_940798.1
8
1693
NR_073610.1
7
1617
XM_005245494.3
XP_005245551.1
6
1861
XM_006711546.2
XP_006711609.1
8
1704
XM_006711547.3
XP_006711610.1
8
2177
XM_006711548.3
XP_006711611.1
7
2059
XM_006711552.2
XP_006711615.1
6
1802
XM_006711553.3
XP_006711616.1
6
1844
XM_011510000.2
XP_011508302.1
7
1967
XM_017002385.1
XP_016857874.1
7
3335
GenBank mRNA
AA310453.1
-
1
49
AF455045.1
-
7
1788
AF455046.1
-
5
1570
AF455047.1
-
7
1532
AK026325.1
-
4
1133
AK054682.1
-
7
1547
AK094346.1
-
7
1586
AK095143.1
-
8
1665
AK095197.1
-
7
1876
AK307441.1
-
6
1397
AL834227.1
-
7
1761
AY424284.1
-
8
1614
BC058509.1
-
8
1577
5770270
BC071623.1
-
3
854
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
steroid hormone mediated signaling pathway
response to steroid hormone
steroid hormone receptor activity
steroid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline