Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290331.1 | NP_001277260.1 | 4 | 327 | ||
NM_024898.3 | NP_079174.2 | 6 | 436 | |||
XM_006722905.2 | XP_006722968.2 | 5 | 306 | |||
XM_006722906.3 | XP_006722969.1 | 6 | 365 | |||
XM_011528318.2 | XP_011526620.1 | 5 | 356 | |||
GenBank mRNA | AK026410.1 | - | 6 | 415 | ||
AK225623.1 | - | 6 | 415 | |||
AK303226.1 | - | 4 | 306 | |||
AK303509.1 | - | 5 | 341 | |||
AL713770.1 | - | 5 | 544 | |||
BC033437.1 | - | 6 | 403 | 5187180 | ||
DC378383.1 | - | 6 | 436 |