Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122841.2 | NP_001116313.1 | 4, 5 | 375 | ||
NM_024919.4 | NP_079195.3 | 4, 5 | 525 | |||
XM_011536137.1 | XP_011534439.1 | 5, 6 | 605 | |||
XM_011536138.1 | XP_011534440.1 | 5, 6 | 421 | |||
XM_011536140.1 | XP_011534442.1 | 4, 5 | 460 | |||
XM_011536141.1 | XP_011534443.1 | 4, 5 | 400 | |||
XM_011536142.2 | XP_011534444.1 | 4, 5 | 438 | |||
XM_011536143.1 | XP_011534445.1 | 4, 5 | 336 | |||
XM_011536145.1 | XP_011534447.1 | 4, 5 | 375 | |||
XM_017011317.1 | XP_016866806.1 | 5, 6 | 605 | |||
GenBank mRNA | AK097179.1 | - | 4, 5 | 525 | ||
AK097579.1 | - | 4, 5 | 375 | |||
AK303194.1 | - | 3, 4 | 179 | |||
AK303407.1 | - | 2, 3 | 233 | |||
AK316478.1 | - | 3, 4 | 179 | |||
BC137144.1 | - | 4, 5 | 483 | 9021141 |