Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185176.1 | NP_001172105.1 | 4 | 760 | ||
NM_001185177.1 | NP_001172106.1 | 11 | 1476 | |||
NM_024922.5 | NP_079198.2 | 11 | 1476 | |||
XM_011522975.2 | XP_011521277.1 | 11 | 1548 | |||
GenBank mRNA | AK000105.1 | - | 11 | 1415 | ||
AK025158.1 | - | 1 | 58 | |||
AK025389.1 | - | 4 | 759 | |||
AK097538.1 | - | 10 | 1918 | |||
AY358609.1 | - | 11 | 1402 | |||
BC053670.1 | - | 11 | 1460 | 6145696 |