Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024928.4 | NP_079204.2 | 2, 3 | 384 | ||
XM_006717976.3 | XP_006718039.1 | 2, 3 | 384 | |||
XM_011540184.2 | XP_011538486.1 | 2, 3 | 384 | |||
XM_017016669.1 | XP_016872158.1 | 1, 2 | 551 | |||
XM_017016670.1 | XP_016872159.1 | 2, 3 | 384 | |||
GenBank mRNA | AK026212.1 | - | 1, 2 | 209 | ||
BC017400.2 | - | 2, 3 | 305 | 3029792 |