Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127383.1 | NP_001120855.1 | 3 | 801 | ||
NM_001256909.1 | NP_001243838.1 | 4 | 672 | |||
NM_024843.3 | NP_079119.3 | 4 | 1010 | |||
GenBank mRNA | AK027115.1 | - | 4 | 852 | ||
AK128495.1 | - | 3 | 680 | |||
AK225459.1 | - | 4 | 852 | |||
AK291731.1 | - | 4 | 860 | |||
AK301485.1 | - | 4 | 672 | |||
AK316588.1 | - | 4 | 889 | |||
AL136693.1 | - | 4 | 893 | |||
AL834483.1 | - | 1 | 173 | |||
BC065290.1 | - | 4 | 845 | 6063145 |