Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA0319L SIRNA Assays ›
Gene Symbol:
KIAA0319L
Gene Name:
KIAA0319 like
Gene Aliases:
AAVR
Chromosome Location:
Chr.1: 35433490 - 35557670 on Build GRCh38
UniGene ID:
Hs.456507
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32023
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA0319L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024874.4
NP_079150.3
17
2879
XM_006710907.1
XP_006710970.1
17
2678
XM_006710909.1
XP_006710972.1
17
2770
XM_006710910.1
XP_006710973.1
16
2322
XM_011542179.2
XP_011540481.1
17
3010
XM_011542180.1
XP_011540482.1
17
2848
XM_017002366.1
XP_016857855.1
17
2863
XM_017002367.1
XP_016857856.1
17
3025
XM_017002368.1
XP_016857857.1
17
2693
XM_017002369.1
XP_016857858.1
17
2785
XM_017002370.1
XP_016857859.1
17
2863
XM_017002371.1
XP_016857860.1
17
2863
XM_017002372.1
XP_016857861.1
17
2770
XM_017002374.1
XP_016857863.1
16
2337
XM_017002375.1
XP_016857864.1
14
1979
XM_017002376.1
XP_016857865.1
14
1964
XM_017002377.1
XP_016857866.1
16
2322
XM_017002378.1
XP_016857867.1
8
1003
GenBank mRNA
AB058740.1
-
12
1577
AF275679.2
-
17
2828
AF289597.1
-
15
2444
AK024287.1
-
13
1642
AK090878.1
-
8
1004
AK314605.1
-
17
2827
AL834315.1
-
13
1713
AY163234.1
-
17
2828
BC001858.2
-
1
41
BC007645.1
-
4
533
BC014530.1
-
17
2811
3843301
BC031672.1
-
12
1573
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline