Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168474.1 | NP_001161946.1 | 13 | 1426 | ||
NM_024885.3 | NP_079161.3 | 13 | 1594 | |||
XM_005262152.1 | XP_005262209.1 | 13 | 1429 | |||
XM_006724664.1 | XP_006724727.1 | 13 | 1596 | |||
GenBank mRNA | AF285595.1 | - | 13 | 1422 | ||
AK026810.1 | - | 11 | 1350 | |||
AK308261.1 | - | 13 | 1596 | |||
BC043391.1 | - | 13 | 1580 | 6154641 |