Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNDIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323606.1 | NP_001310535.1 | 5 | 2528 | ||
NM_001323607.1 | NP_001310536.1 | 4 | 1204 | |||
NM_024893.2 | NP_079169.1 | 4 | 1639 | |||
XM_017028065.1 | XP_016883554.1 | 5 | 2000 | |||
XM_017028066.1 | XP_016883555.1 | 4 | 1357 | |||
XM_017028067.1 | XP_016883556.1 | 4 | 1263 | |||
XM_017028068.1 | XP_016883557.1 | 4 | 2224 | |||
GenBank mRNA | AK024282.1 | - | 4 | 1639 | ||
BC030637.1 | - | 4 | 1141 | 4821088 |