Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271989.1 | NP_001258918.1 | 6, 7 | 945 | ||
NM_024899.3 | NP_079175.2 | 7, 8 | 1170 | |||
NR_073537.1 | 7, 8 | 1165 | ||||
XM_005258149.4 | XP_005258206.1 | 7, 8 | 1149 | |||
XM_017025981.1 | XP_016881470.1 | 7, 8 | 1149 | |||
XM_017025982.1 | XP_016881471.1 | 7, 8 | 1230 | |||
XM_017025983.1 | XP_016881472.1 | 6, 7 | 1020 | |||
XM_017025984.1 | XP_016881473.1 | 7, 8 | 1132 | |||
XM_017025985.1 | XP_016881474.1 | 7, 8 | 1098 | |||
XM_017025986.1 | XP_016881475.1 | 6, 7 | 907 | |||
XM_017025987.1 | XP_016881476.1 | 6, 7 | 874 | |||
GenBank mRNA | AK301964.1 | - | 6, 7 | 907 | ||
AK303117.1 | - | 6, 7 | 892 | |||
AL833727.1 | - | 8, 9 | 1162 | |||
BC026307.1 | - | 7, 8 | 1097 | 4792211 | ||
BC033026.1 | - | 7, 8 | 1032 |