Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271833.1 | NP_001258762.1 | 11, 12 | 1523 | ||
NM_024901.4 | NP_079177.2 | 11, 12 | 1591 | |||
XM_006710921.2 | XP_006710984.2 | 12, 13 | 1545 | |||
XM_006710922.1 | XP_006710985.1 | 11, 12 | 1749 | |||
XM_006710923.1 | XP_006710986.1 | 11, 12 | 1737 | |||
XM_006710924.1 | XP_006710987.1 | 11, 12 | 1514 | |||
XM_011542187.1 | XP_011540489.1 | 12, 13 | 1556 | |||
XM_011542189.1 | XP_011540491.1 | 11, 12 | 1764 | |||
XM_011542190.2 | XP_011540492.1 | 12, 13 | 1424 | |||
XM_017002386.1 | XP_016857875.1 | 11, 12 | 1300 | |||
XM_017002387.1 | XP_016857876.1 | 11, 12 | 1316 | |||
XM_017002388.1 | XP_016857877.1 | 11, 12 | 1550 | |||
GenBank mRNA | AK026110.1 | - | 11, 12 | 1394 | ||
AK301583.1 | - | 10, 11 | 1387 | |||
BC004557.2 | - | 11, 12 | 1547 | 3957641 |